Gene Transcripts

Gene Symbol
Trp53inp1
Entrez Gene ID
60599
Gene Name
transformation related protein 53 inducible nuclear protein 1
Gene Aliases
2700057G22Rik, SIP, SIP18, SIP27, Stinp, Teap
Chromosome Location
Chr. 4: 11156441 - 11174377 on Build GRCm38
UniGene ID
Mm.393018
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199105.1 NP_001186034.1 3 578
NM_021897.3 NP_068697.1 3 578
GenBank mRNA AJ131776.1 - 3 579
AK054214.1 - 3 575
AK142321.1 - 3 574
AY034611.1 - 3 579
AY034612.1 - 3 579
BC132124.1 - 2 480 40130401
BC138711.1 - 2 480 8861733
BC145396.1 - 2 479

View Full Product Details