Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000686.4 | NP_000677.2 | 3 | 446 | |
XM_011537533.1 | XP_011535835.1 | 2 | 344 | ||
GenBank mRNA | AK313927.1 | - | 3 | 448 | |
BC095504.1 | - | 1 | 707 | 6971944 | |
CR541969.1 | - | 1 | 239 | ||
CR738498.1 | - | 3 | 414 | ||
DA538369.1 | - | 3 | 448 | ||
U16957.1 | - | 3 | 379 |