Gene Transcripts

Gene Symbol
Emc3
Entrez Gene ID
66087
Gene Name
ER membrane protein complex subunit 3
Gene Aliases
0610039A15Rik, AI225901, AW260416, Pob, Tmem111
Chromosome Location
Chr. 6: 113514887 - 113531638 on Build GRCm38
UniGene ID
Mm.275989
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175101.3 NP_780310.1 5 581
GenBank mRNA AK020058.1 - 5 552
AK075595.1 - 5 582
AK167819.1 - 5 430
AK200024.1 - 2 34
AK210449.1 - 5 291
AK218757.1 - 2 34
BC004641.1 - 5 577 3157147

View Full Product Details