Gene Transcripts

Gene Symbol
Letm1
Entrez Gene ID
56384
Gene Name
leucine zipper-EF-hand containing transmembrane protein 1
Gene Aliases
Chromosome Location
Chr. 5: 33739675 - 33782704 on Build GRCm38
UniGene ID
Mm.260538
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019694.1 NP_062668.1 14 3335
GenBank mRNA AF061026.1 - 14 3335
AK163499.1 - 1 697
BC061690.1 - 14 3334

View Full Product Details