Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019786.4 | NP_062760.3 | 6 | 746 | |
GenBank mRNA | AF145705.1 | - | 5 | 598 | |
AF191839.1 | - | 5 | 625 | ||
AK004649.2 | - | 6 | 700 | ||
AK083214.1 | - | 6 | 693 | ||
AK168193.1 | - | 6 | 728 | ||
BC129910.1 | - | 5 | 658 | 40111752 | |
BC129911.1 | - | 5 | 658 | 40111754 |