Gene Transcripts

Gene Symbol
Trim17
Entrez Gene ID
56631
Gene Name
tripartite motif-containing 17
Gene Aliases
Rnf16, terf
Chromosome Location
Chr. 11: 58955310 - 58971737 on Build GRCm38
UniGene ID
Mm.179733
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031172.2 NP_112449.1 3 543
GenBank mRNA AK077212.1 - 3 524
BC055112.1 - 3 543 6707359

View Full Product Details