Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_053014.3 | NP_443747.2 | 3 | 263 | |
GenBank mRNA | AB377215.1 | - | 1 | 79 | |
AK008965.1 | - | 3 | 421 | ||
AK015906.1 | - | 2 | 191 | ||
AK030607.1 | - | 1 | 55 | ||
AK075715.1 | - | 3 | 263 | ||
AK076414.1 | - | 2 | 232 | ||
AK159110.1 | - | 2 | 503 | ||
AY167588.1 | - | 1 | 101 | ||
BC052382.1 | - | 2 | 154 | 5703571 | |
BC058519.1 | - | 2 | 153 | 5703571 |