Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163643.1 | NP_001157115.1 | 6 | 1120 | |
NM_009582.4 | NP_033608.3 | 4 | 944 | ||
GenBank mRNA | AK029882.1 | - | 4 | 753 | |
AK034374.1 | - | 6 | 936 | ||
AK166435.1 | - | 6 | 1040 | ||
BC047158.1 | - | 3 | 738 | 6494216 | |
BC057572.1 | - | 6 | 944 | 6401321 | |
U14636.1 | - | 4 | 776 | ||
U23789.1 | - | 3 | 734 |