Gene Transcripts

Gene Symbol
Scn11a
Entrez Gene ID
24046
Gene Name
sodium channel, voltage-gated, type XI, alpha
Gene Aliases
NSS2, NaN, NaT, NaV1.9, SNS2
Chromosome Location
Chr. 9: 119753730 - 119825456 on Build GRCm38
UniGene ID
Mm.469833
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011887.3 NP_036017.3 9, 10 1143
GenBank mRNA AB031389.1 - 9, 10 1201
AF118044.1 - 9, 10 1143

View Full Product Details