Gene Transcripts

Gene Symbol
Vax2
Entrez Gene ID
24113
Gene Name
ventral anterior homeobox 2
Gene Aliases
Dres93
Chromosome Location
Chr. 6: 83711264 - 83739229 on Build GRCm38
UniGene ID
Mm.307165
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011912.3 NP_036042.1 3 1072
GenBank mRNA AF028715.1 - 3 1046
BC090635.1 - 3 1069 30942578
Y17792.1 - 3 1072
Y18247.1 - 1 160

View Full Product Details