Gene Transcripts
- Gene Symbol
- RAB31
- Entrez Gene ID
- 11031
- Gene Name
- RAB31, member RAS oncogene family
- Gene Aliases
- Rab22B
- Chromosome Location
- Chr. 18: 9707971 - 9862556 on Build GRCh38
- UniGene ID
- Hs.744887
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006868.3 |
NP_006859.2
|
3, 4 |
378 |
|
XM_017025528.1 |
XP_016881017.1
|
4, 5 |
760 |
|
XM_017025529.1 |
XP_016881018.1
|
4, 5 |
748 |
|
GenBank mRNA |
AF183421.1 |
- |
3, 4 |
331 |
|
AF234995.1 |
- |
3, 4 |
377 |
|
AF498957.1 |
- |
3, 4 |
200 |
|
AK223505.1 |
- |
3, 4 |
242 |
|
AK314809.1 |
- |
3, 4 |
336 |
|
BC001148.1 |
- |
3, 4 |
300 |
3534853 |
BT020027.1 |
- |
3, 4 |
200 |
|
BT020028.1 |
- |
3, 4 |
200 |
|
BU944638.1 |
- |
3, 4 |
444 |
|
CR407659.1 |
- |
3, 4 |
200 |
|
U57091.1 |
- |
3, 4 |
245 |
|
U59877.1 |
- |
3, 4 |
260 |
|
View Full Product Details