Gene Transcripts

Gene Symbol
Crip2
Entrez Gene ID
338401
Gene Name
cysteine-rich protein 2
Gene Aliases
CRP2
Chromosome Location
Chr. 6: 137953545 - 137958491 on Build Rnor_6.0
UniGene ID
Rn.4267
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022501.1 NP_071946.1 2, 3 254
XM_008764943.1 XP_008763165.1 2, 3 518
GenBank mRNA BC061774.1 - 2, 3 223
D17512.1 - 2, 3 254
FQ226314.1 - 2, 3 262
FQ227529.1 - 2, 3 264
FQ230331.1 - 2, 3 264
FQ233584.1 - 2, 3 268

View Full Product Details