Gene Transcripts

Gene Symbol
Dynlt1
Entrez Gene ID
83462
Gene Name
dynein light chain Tctex-type 1
Gene Aliases
AGS2, Tctex1
Chromosome Location
Chr. 1: 47206892 - 47213782 on Build Rnor_6.0
UniGene ID
Rn.29901
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031318.1 NP_112608.1 5 462
XM_006227901.2 XP_006227963.1 4 933
GenBank mRNA AJ131437.1 - 5 462
BC166879.1 - 5 494 6919674

View Full Product Details