Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031318.1 | NP_112608.1 | 3 | 150 | |
XM_006227901.2 | XP_006227963.1 | 3 | 215 | ||
GenBank mRNA | AB010119.1 | - | 3 | 139 | |
AJ131437.1 | - | 3 | 150 | ||
BC166879.1 | - | 3 | 182 | 6919674 |