Gene Transcripts

Gene Symbol
Sh3bp5
Entrez Gene ID
117186
Gene Name
SH3-domain binding protein 5 (BTK-associated)
Gene Aliases
SH3BP-5, Sab, Vesp18
Chromosome Location
Chr. 16: 7473362 - 7538201 on Build Rnor_6.0
UniGene ID
Rn.163321
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_054011.1 NP_446463.2 2 183
GenBank mRNA AB027562.1 - 2 183
FQ210473.1 - 2 310

View Full Product Details