Gene Transcripts

Gene Symbol
Dad1
Entrez Gene ID
192275
Gene Name
defender against cell death 1
Gene Aliases
Chromosome Location
Chr. 15: 32868136 - 32887933 on Build Rnor_6.0
UniGene ID
Rn.7262
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138910.3 NP_620265.1 3 508
GenBank mRNA BC061530.1 - 3 512
FQ222612.1 - 3 508
FQ223332.1 - 3 517

View Full Product Details