Gene Transcripts

Gene Symbol
Slc25a25
Entrez Gene ID
246771
Gene Name
solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 25
Gene Aliases
Mcsc, Pcscl
Chromosome Location
Chr. 3: 11442396 - 11476186 on Build Rnor_6.0
UniGene ID
Rn.17644
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145677.1 NP_663710.1 1, 2 264
XM_006233935.2 XP_006233997.1 1, 2 1601
GenBank mRNA AY043169.1 - 1, 2 264
FQ211394.1 - 1, 2 280

View Full Product Details