Gene Transcripts

Gene Symbol
Nek6
Entrez Gene ID
360161
Gene Name
NIMA-related kinase 6
Gene Aliases
Chromosome Location
Chr. 3: 22811927 - 22888598 on Build Rnor_6.0
UniGene ID
Rn.18744
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001277232.1 NP_001264161.1 4 345
NM_182953.2 NP_891998.1 4 397
XM_006234100.1 XP_006234162.1 4 420
GenBank mRNA AY334013.1 - 3 294
BC160820.1 - 3 421 7459041

View Full Product Details