Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007115.3 | NP_009046.2 | 6 | 798 | |
GenBank mRNA | AF086484.1 | - | 6 | 650 | |
AJ419936.1 | - | 6 | 723 | ||
AJ421518.1 | - | 6 | 723 | ||
BC030205.1 | - | 6 | 767 | 5208773 | |
M31165.1 | - | 6 | 791 |