Gene Transcripts

Gene Symbol
Ly6i
Entrez Gene ID
246138
Gene Name
lymphocyte antigen 6 complex, locus I
Gene Aliases
Ly6b
Chromosome Location
Chr. 7: 116419573 - 116425017 on Build Rnor_6.0
UniGene ID
Rn.39703
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139257.1 NP_640350.1 4 583
XM_006241765.2 XP_006241827.1 570
XM_006241766.2 XP_006241828.1 5 710
XM_008765545.1 XP_008763767.1 5 697
GenBank mRNA M30689.1 - 4 583

Gene Symbol
Ly6al
Entrez Gene ID
362935
Gene Name
lymphocyte antigen 6 complex, locus A-like
Gene Aliases
Ly6a
Chromosome Location
Chr. 7: 116549110 - 116561263 on Build Rnor_6.0
UniGene ID
Rn.203167
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128099.1 NP_001121571.1 3 481

Gene Symbol
RGD1565410
Entrez Gene ID
503162
Gene Name
similar to Ly6-C antigen gene
Gene Aliases
Chromosome Location
Chr. 7: 116501659 - 116507113 on Build Rnor_6.0
UniGene ID
Rn.215825
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113792.1 NP_001107264.1 637
GenBank mRNA BC158674.1 - 637

View Full Product Details