Gene Transcripts

Gene Symbol
CTRC
Entrez Gene ID
11330
Gene Name
chymotrypsin C
Gene Aliases
CLCR, ELA4
Chromosome Location
Chr. 1: 15438442 - 15449247 on Build GRCh38
UniGene ID
Hs.631869
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007272.2 NP_009203.2 4 323
XM_011540550.1 XP_011538852.1 4 323
GenBank mRNA AK289447.1 - 4 322
BC015118.1 - 4 329 3950170
BC059779.1 - 4 302
CR749296.1 - 2 133
S82198.1 - 4 300
Y13697.1 - 3 252

View Full Product Details