Gene Transcripts

Gene Symbol
Aqp11
Entrez Gene ID
286758
Gene Name
aquaporin 11
Gene Aliases
Chromosome Location
Chr. 1: 162703394 - 162713560 on Build Rnor_6.0
UniGene ID
Rn.20144
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173105.1 NP_775128.1 1 661
GenBank mRNA AB023644.1 - 1 661
BC078694.1 - 1 688 7111950

View Full Product Details