Gene Transcripts
- Gene Symbol
- Myh6
- Entrez Gene ID
- 29556
- Gene Name
- myosin, heavy chain 6, cardiac muscle, alpha
- Gene Aliases
- Myhca
- Chromosome Location
- Chr. 15: 33605653 - 33629592 on Build Rnor_6.0
- UniGene ID
- Rn.54399
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017239.2 |
NP_058935.2
|
6 |
666 |
|
GenBank mRNA |
X15938.1 |
- |
6 |
663 |
|
View Full Product Details