Gene Transcripts

Gene Symbol
Ubd
Entrez Gene ID
29168
Gene Name
ubiquitin D
Gene Aliases
Chromosome Location
Chr. 20: 1876175 - 1878126 on Build Rnor_6.0
UniGene ID
Rn.18473
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053299.2 NP_445751.2 2 469
GenBank mRNA AJ312394.1 - 2 471

View Full Product Details