Gene Transcripts

Gene Symbol
Amacr
Entrez Gene ID
25284
Gene Name
alpha-methylacyl-CoA racemase
Gene Aliases
Da1-8, Marc1
Chromosome Location
Chr. 2: 60949276 - 60961342 on Build Rnor_6.0
UniGene ID
Rn.2590
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012816.2 NP_036948.2 5 785
GenBank mRNA AY325250.1 - 1034
BC078885.1 - 5 753 7110324
BC091163.1 - 5 754 7130642
BC127505.1 - 5 774 7325347
U89905.1 - 5 773
Y08172.1 - 5 763

View Full Product Details