Gene Transcripts
- Gene Symbol
- Alb
- Entrez Gene ID
- 24186
- Gene Name
- albumin
- Gene Aliases
- Alb1, Albza
- Chromosome Location
- Chr. 14: 19176275 - 19191793 on Build Rnor_6.0
- UniGene ID
- Rn.202968
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_134326.2 |
NP_599153.2
|
2, 3 |
146 |
|
GenBank mRNA |
BC085359.1 |
- |
2, 3 |
146 |
|
FQ209504.1 |
- |
2, 3 |
168 |
|
FQ209784.1 |
- |
2, 3 |
165 |
|
FQ210189.1 |
- |
2, 3 |
165 |
|
FQ210310.1 |
- |
2, 3 |
164 |
|
FQ210436.1 |
- |
2, 3 |
165 |
|
FQ211062.1 |
- |
2, 3 |
169 |
|
FQ211231.1 |
- |
2, 3 |
165 |
|
FQ211307.1 |
- |
2, 3 |
165 |
|
FQ211332.1 |
- |
2, 3 |
165 |
|
FQ218069.1 |
- |
2, 3 |
168 |
|
FQ218125.1 |
- |
2, 3 |
165 |
|
FQ219734.1 |
- |
2, 3 |
161 |
|
V01222.1 |
- |
2, 3 |
131 |
|
View Full Product Details