Gene Transcripts

Gene Symbol
Zg16
Entrez Gene ID
171449
Gene Name
zymogen granule protein 16
Gene Aliases
Zg-16p, Zg16p
Chromosome Location
Chr. 1: 198483800 - 198486157 on Build Rnor_6.0
UniGene ID
Rn.10439
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134409.2 NP_599236.2 3 147
GenBank mRNA AB095177.1 - 3 147
CF249177.1 - 2 119
Z30584.1 - 3 176

View Full Product Details