Gene Transcripts

Gene Symbol
CFAP20
Entrez Gene ID
29105
Gene Name
cilia and flagella associated protein 20
Gene Aliases
C16orf80, EVORF, GTL3, fSAP23
Chromosome Location
Chr. 16: 58113588 - 58129425 on Build GRCh38
UniGene ID
Hs.532755
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013242.2 NP_037374.1 1, 2 369
NR_136632.1 1, 2 402
GenBank mRNA AF087845.1 - 1, 2 349
AF093680.1 - 1, 2 348
AK297819.1 - 1, 2 324
AV751497.1 - 1, 2 369
BC005152.2 - 1, 2 338 3528301
CN364433.1 - 1, 2 329
HY042096.1 - 1, 2 403

View Full Product Details