Gene Transcripts
- Gene Symbol
- TAC3
- Entrez Gene ID
- 6866
- Gene Name
- tachykinin 3
- Gene Aliases
- HH10, NKB, NKNB, PRO1155, ZNEUROK1
- Chromosome Location
- Chr. 12: 57009997 - 57016560 on Build GRCh38
- UniGene ID
- Hs.9730
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001178054.1 |
NP_001171525.1
|
2, 3 |
303 |
|
NM_013251.3 |
NP_037383.1
|
2, 3 |
303 |
|
NR_033654.1 |
|
2, 3 |
303 |
|
NR_135164.1 |
|
2, 3 |
303 |
|
NR_135165.1 |
|
2, 3 |
303 |
|
NR_135166.1 |
|
2, 3 |
303 |
|
GenBank mRNA |
AF186112.1 |
- |
2, 3 |
246 |
|
AF216586.1 |
- |
2, 3 |
233 |
|
AF537113.1 |
- |
2, 3 |
263 |
|
AF537114.1 |
- |
2, 3 |
263 |
|
AF537115.1 |
- |
2, 3 |
263 |
|
AF537116.1 |
- |
2, 3 |
263 |
|
AF537117.1 |
- |
2, 3 |
263 |
|
AF537118.1 |
- |
2, 3 |
263 |
|
AF537119.1 |
- |
2, 3 |
263 |
|
AF537120.1 |
- |
2, 3 |
263 |
|
AF537121.1 |
- |
2, 3 |
263 |
|
AX147032.1 |
- |
2, 3 |
149 |
|
AY358679.1 |
- |
2, 3 |
281 |
|
BC032145.2 |
- |
2, 3 |
284 |
4897300 |
BP212420.1 |
- |
2, 3 |
303 |
|
CR457193.1 |
- |
1, 2 |
124 |
|
View Full Product Details