Gene Transcripts

Gene Symbol
Lppr1
Entrez Gene ID
298062
Gene Name
lipid phosphate phosphatase-related protein type 1
Gene Aliases
Prg-3
Chromosome Location
Chr. 5: 67852817 - 67985310 on Build Rnor_6.0
UniGene ID
Rn.215023
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201271.1 NP_958428.1 2 222
GenBank mRNA AY299399.1 - 2 222

View Full Product Details