Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_178422.5 | NP_848509.1 | 2 | 2248 | |
XM_005245745.3 | XP_005245802.1 | 3 | 2070 | ||
XM_005245746.3 | XP_005245803.1 | 2 | 1986 | ||
XM_011540861.1 | XP_011539163.1 | 3 | 1893 | ||
XM_011540862.2 | XP_011539164.1 | 4 | 2159 | ||
GenBank mRNA | BC034015.1 | - | 2 | 2248 |