Gene Transcripts
- Gene Symbol
- SACM1L
- Entrez Gene ID
- 22908
- Gene Name
- SAC1 suppressor of actin mutations 1-like (yeast)
- Gene Aliases
- SAC1
- Chromosome Location
- Chr. 3: 45689241 - 45745425 on Build GRCh38
- UniGene ID
- Hs.156509
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001319072.1 |
NP_001306001.1
|
4, 5 |
587 |
|
NM_001319073.1 |
NP_001306002.1
|
6, 7 |
755 |
|
NM_014016.4 |
NP_054735.3
|
5, 6 |
715 |
|
XM_011533500.2 |
XP_011531802.1
|
5, 6 |
506 |
|
GenBank mRNA |
AB020658.1 |
- |
5, 6 |
559 |
|
AK291142.1 |
- |
5, 6 |
599 |
|
AK296421.1 |
- |
4, 5 |
369 |
|
AK301248.1 |
- |
6, 7 |
595 |
|
AL136831.1 |
- |
5, 6 |
505 |
|
BC016559.1 |
- |
5, 6 |
511 |
3049075 |
CR749564.1 |
- |
5, 6 |
1661 |
|
View Full Product Details