Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014508.2 | NP_055323.2 | 4 | 1084 | |
GenBank mRNA | AB208979.1 | - | 4 | 1049 | |
AF165520.1 | - | 4 | 1089 | ||
BC011739.2 | - | 4 | 1084 | 4331363 | |
BC021080.1 | - | 4 | 1072 |