Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001292038.1 | NP_001278967.1 | 13 | 2048 | |
NM_015274.2 | NP_056089.1 | 13 | 2201 | ||
GenBank mRNA | AB023152.2 | - | 13 | 2160 | |
AK025803.1 | - | 5 | 829 | ||
AK075246.1 | - | 1 | 139 | ||
AK301473.1 | - | 13 | 2033 | ||
AL832232.1 | - | 13 | 2171 | ||
AL833071.2 | - | 13 | 2174 | ||
BC033307.1 | - | 13 | 2170 | 4932398 | |
BC036836.1 | - | 3 | 383 | ||
BC094773.1 | - | 13 | 2201 | 30531368 |