Gene Transcripts

Gene Symbol
MSTO2P
Entrez Gene ID
100129405
Gene Name
misato family member 2, pseudogene
Gene Aliases
MSTO2
Chromosome Location
Chr. 1: 155745768 - 155750688 on Build GRCh38
UniGene ID
Hs.656361
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024117.2 13 1675
GenBank mRNA BC099909.1 - 13 1628
CX784525.1 - 2 228

Gene Symbol
MSTO1
Entrez Gene ID
55154
Gene Name
misato 1, mitochondrial distribution and morphology regulator
Gene Aliases
LST005, MST
Chromosome Location
Chr. 1: 155610170 - 155614967 on Build GRCh38
UniGene ID
Hs.656547
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256532.1 NP_001243461.1 14 1784
NM_001256533.1 NP_001243462.1 14 1751
NM_018116.3 NP_060586.2 14 1787
NR_046292.1 14 1876
NR_046293.1 14 1818
NR_046294.1 12 1810
NR_046295.1 13 1711
XM_006711421.1 XP_006711484.1 13 1746
XM_006711423.2 XP_006711486.1 14 1795
XM_011509695.1 XP_011507997.1 14 1807
XM_011509696.1 XP_011507998.1 14 1712
XM_011509697.1 XP_011507999.1 14 1782
XM_011509698.1 XP_011508000.1 14 1779
XM_011509699.1 XP_011508001.1 14 1770
XM_011509700.1 XP_011508002.1 14 1749
XM_011509701.1 XP_011508003.1 14 1746
XM_011509702.1 XP_011508004.1 13 1592
XM_011509703.1 XP_011508005.1 13 2387
XM_011509704.1 XP_011508006.1 14 2310
XM_011509705.1 XP_011508007.1 13 2324
XM_011509706.1 XP_011508008.1 14 1744
XM_011509707.1 XP_011508009.1 14 2252
XM_011509708.1 XP_011508010.1 14 2185
XM_017001603.1 XP_016857092.1 14 1808
XM_017001604.1 XP_016857093.1 14 1746
XM_017001605.1 XP_016857094.1 14 2277
XM_017001606.1 XP_016857095.1 14 2188
XM_017001607.1 XP_016857096.1 14 2326
XM_017001608.1 XP_016857097.1 13 1773
XM_017001609.1 XP_016857098.1 14 1792
XM_017001610.1 XP_016857099.1 14 1783
XM_017001611.1 XP_016857100.1 14 2314
XM_017001612.1 XP_016857101.1 13 1725
XM_017001613.1 XP_016857102.1 13 1653
XM_017001614.1 XP_016857103.1 14 1820
XM_017001615.1 XP_016857104.1 14 1882
XM_017001616.1 XP_016857105.1 9 1336
GenBank mRNA AF111708.1 - 8 1863
AF272833.1 - 14 1665
AK001366.1 - 14 1716
AK056128.1 - 12 1739
AK092950.1 - 13 1578
AK222863.1 - 14 1710
AK290610.1 - 14 1759
AK297140.1 - 14 1805
AK309573.1 - 14 1739
AY334564.1 - 13 1827
BC002535.2 - 14 1732 3140498
BC070067.1 - 10 2523
BX537684.1 - 13 1663
CR749791.1 - 14 1677
CR936872.1 - 10 2755

View Full Product Details