Gene Transcripts

Gene Symbol
UTP20
Entrez Gene ID
27340
Gene Name
UTP20, small subunit processome component
Gene Aliases
1A6/DRIM, DRIM
Chromosome Location
Chr. 12: 101280127 - 101386619 on Build GRCh38
UniGene ID
Hs.295732
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014503.2 NP_055318.2 2, 3 290
GenBank mRNA AJ006778.1 - 2, 3 278
AK307934.1 - 2, 3 290
BX957203.1 - 2, 3 309

View Full Product Details