Gene Transcripts

Gene Symbol
LCE1E
Entrez Gene ID
353135
Gene Name
late cornified envelope 1E
Gene Aliases
LEP5
Chromosome Location
Chr. 1: 152786277 - 152788426 on Build GRCh38
UniGene ID
Hs.250236
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178353.1 NP_848130.1 2 1089
GenBank mRNA BC038391.1 - 2 1089

View Full Product Details