Gene Transcripts

Gene Symbol
OR2T4
Entrez Gene ID
127074
Gene Name
olfactory receptor family 2 subfamily T member 4
Gene Aliases
OR1-60, OR2T4Q
Chromosome Location
Chr. 1: 248361581 - 248362627 on Build GRCh38
UniGene ID
Hs.553586
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004696.1 NP_001004696.1 1 1029

View Full Product Details