Gene Transcripts

Gene Symbol
EIF5A
Entrez Gene ID
1984
Gene Name
eukaryotic translation initiation factor 5A
Gene Aliases
EIF-5A, EIF5A1, eIF5AI
Chromosome Location
Chr. 17: 7306999 - 7312463 on Build GRCh38
UniGene ID
Hs.534314
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143760.1 NP_001137232.1 6 711
NM_001143761.1 NP_001137233.1 6 739
NM_001143762.1 NP_001137234.1 6 795
NM_001970.4 NP_001961.1 6 810
XM_005256509.2 XP_005256566.1 6 730
GenBank mRNA AF087992.1 - 5 524
AK222585.1 - 6 609
AK292615.1 - 6 647
AY129319.1 - 6 711
AY129320.1 - 6 798
AY129321.1 - 6 723
AY129322.1 - 6 767
BC000751.2 - 6 697 2961296
BC001832.2 - 6 697 2961296
BC030160.2 - 6 685 5022012
BC080196.1 - 6 680 6528111
BC085015.1 - 5 605 6575132
BC107779.1 - 6 689 6495558

Gene Symbol
EIF5AL1
Entrez Gene ID
143244
Gene Name
eukaryotic translation initiation factor 5A-like 1
Gene Aliases
EIF5AP1, bA342M3.3
Chromosome Location
Chr. 10: 79512601 - 79516440 on Build GRCh38
UniGene ID
Hs.104825
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099692.1 NP_001093162.1 1 622
GenBank mRNA BC070048.1 - 1 622

View Full Product Details