Gene Transcripts

Gene Symbol
Clec4a2
Entrez Gene ID
297584
Gene Name
C-type lectin domain family 4, member A2
Gene Aliases
Dcir2
Chromosome Location
Chr. 4: 155793059 - 155867512 on Build Rnor_6.0
UniGene ID
Rn.129074
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005880.1 NP_001005880.1 5 563
GenBank mRNA AY397670.1 - 5 563
AY494061.1 - 5 564
AY581052.1 - 5 563

View Full Product Details