Gene Transcripts

Gene Symbol
Tob2
Entrez Gene ID
315159
Gene Name
transducer of ERBB2, 2
Gene Aliases
Chromosome Location
Chr. 7: 123079520 - 123088240 on Build Rnor_6.0
UniGene ID
Rn.18247
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007146.1 NP_001007147.1 2 1837
GenBank mRNA AY724519.1 - 2 2040
BC085117.1 - 2 1876 7107186

View Full Product Details