Gene Transcripts

Gene Symbol
Snf8
Entrez Gene ID
287645
Gene Name
SNF8, ESCRT-II complex subunit
Gene Aliases
D11moh34, RGD1310144
Chromosome Location
Chr. 10: 83856331 - 83868621 on Build Rnor_6.0
UniGene ID
Rn.162167
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007804.1 NP_001007805.1 3 142
XM_006247185.1 XP_006247247.1 3 275
GenBank mRNA BC086364.1 - 3 185

View Full Product Details