Gene Transcripts

Gene Symbol
TRIM72
Entrez Gene ID
493829
Gene Name
tripartite motif containing 72
Gene Aliases
MG53
Chromosome Location
Chr. 16: 31214021 - 31225656 on Build GRCh38
UniGene ID
Hs.661859
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008274.3 NP_001008275.2 7 1770
XM_017023254.1 XP_016878743.1 6 1306
GenBank mRNA AK090695.1 - 6 1215
AK131485.1 - 7 1715

View Full Product Details