Gene Transcripts

Gene Symbol
Pgrmc2
Entrez Gene ID
361940
Gene Name
progesterone receptor membrane component 2
Gene Aliases
Chromosome Location
Chr. 2: 127986106 - 128002005 on Build Rnor_6.0
UniGene ID
Rn.7685
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008374.1 NP_001008375.1 2 510
GenBank mRNA BC083571.1 - 2 510

View Full Product Details