Gene Transcripts

Gene Symbol
Scn4b
Entrez Gene ID
315611
Gene Name
sodium channel, voltage-gated, type IV, beta subunit
Gene Aliases
Chromosome Location
Chr. 8: 49441106 - 49456279 on Build Rnor_6.0
UniGene ID
Rn.22230
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008880.1 NP_001008880.1 2 124

View Full Product Details