Gene Transcripts

Gene Symbol
PLEKHH1
Entrez Gene ID
57475
Gene Name
pleckstrin homology, MyTH4 and FERM domain containing H1
Gene Aliases
Chromosome Location
Chr. 14: 67533155 - 67589766 on Build GRCh38
UniGene ID
Hs.594236
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020715.2 NP_065766.1 29 5816
XM_011537009.1 XP_011535311.1 29 5992
XM_011537015.2 XP_011535317.1 22 6047
XM_017021499.1 XP_016876988.1 28 5973
XM_017021500.1 XP_016876989.1 29 5908
XM_017021501.1 XP_016876990.1 28 5889
XM_017021502.1 XP_016876991.1 22 5769
XM_017021503.1 XP_016876992.1 22 5129
XM_017021504.1 XP_016876993.1 24 5271
GenBank mRNA AB033026.1 - 29 5792
AK123262.1 - 2 2280
AK124787.1 - 6 2873
BC014159.2 - 6 2275
BC037820.1 - 1 1722
BC146787.1 - 29 5792 9007246
BX648137.1 - 6 5148
BX648464.1 - 20 4929

View Full Product Details