Gene Transcripts

Gene Symbol
ZYG11A
Entrez Gene ID
440590
Gene Name
zyg-11 family member A, cell cycle regulator
Gene Aliases
ZYG11
Chromosome Location
Chr. 1: 52842511 - 52894575 on Build GRCh38
UniGene ID
Hs.658458
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004339.2 NP_001004339.2 12, 13 2381
NM_001307931.1 NP_001294860.1 11, 12 1629
XM_011541480.2 XP_011539782.1 11, 12 2111
XM_011541481.2 XP_011539783.1 11, 12 1979
XM_011541482.2 XP_011539784.1 11, 12 1961
XM_011541483.2 XP_011539785.1 10, 11 1916
GenBank mRNA AY271826.1 - 11, 12 1620

View Full Product Details