Gene Transcripts

Gene Symbol
Letm1
Entrez Gene ID
305457
Gene Name
leucine zipper-EF-hand containing transmembrane protein 1
Gene Aliases
Chromosome Location
Chr. 14: 82227790 - 82267298 on Build Rnor_6.0
UniGene ID
Rn.162290
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005884.1 NP_001005884.1 10, 11 1804
GenBank mRNA BC083642.1 - 10, 11 1804

View Full Product Details