Gene Transcripts

Gene Symbol
Zfp85
Entrez Gene ID
22746
Gene Name
zinc finger protein 85
Gene Aliases
KRAB19, Rslcan2, Zfp71, Zfp85-rs1
Chromosome Location
Chr. 13: 67744464 - 67755134 on Build GRCm38
UniGene ID
Mm.288396
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001130.2 NP_001001130.1 4 1633
GenBank mRNA BC055337.1 - 4 1633
BC055756.1 - 4 1633 6413269

View Full Product Details