Gene Transcripts

Gene Symbol
Fndc5
Entrez Gene ID
384061
Gene Name
fibronectin type III domain containing 5
Gene Aliases
1500001L03Rik, AI836596, C87088, PeP, Pxp
Chromosome Location
Chr. 4: 129136922 - 129144593 on Build GRCm38
UniGene ID
Mm.44075
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027402.3 NP_081678.1 6 2075
GenBank mRNA AA003587.1 - 1 191
AJ401376.1 - 6 2075
AK137327.1 - 1 759
CX222088.1 - 1 620

View Full Product Details